Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tenc1
Entrez Gene ID
100752687
Gene Name
tensin like C1 domain containing phosphatase (tensin 2)
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617247: 55755 - 72176 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655053.1 XP_007653243.1 8 - 9 1304 113

View Full Product Details