Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnfsf13b
Entrez Gene ID
100757266
Gene Name
tumor necrosis factor (ligand) superfamily, member 13b
Gene Aliases
I79_001029
Chromosome Location
Chr.Un|NW_003613600: 4007761 - 4033066 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003495769.2 XP_003495817.2 5 - 6 718 86
XM_007643890.1 XP_007642080.1 4 - 5 661 86

View Full Product Details