Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crygn
Entrez Gene ID
100759221
Gene Name
crystallin, gamma N
Gene Aliases
I79_007676
Chromosome Location
Chr.Un|NW_003613842: 752396 - 760580 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501285.2 XP_003501333.1 2 - 3 273 78

View Full Product Details