Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cetn3
Entrez Gene ID
100765724
Gene Name
centrin, EF-hand protein, 3
Gene Aliases
Centrin-3
Chromosome Location
Chr.Un|NW_003614058: 503779 - 519257 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003504389.2 XP_003504437.1 2 - 3 266 70
GenBank mRNA JI892957.1 - 2 - 3 260 70
JP048562.1 - 2 - 3 260 70

View Full Product Details