Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gabarapl2
Entrez Gene ID
100767407
Gene Name
GABA(A) receptor-associated protein-like 2
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614840: 135768 - 154923 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003510161.2 XP_003510209.1 4 - 5 488 105
GenBank mRNA JI889706.1 - 3 - 4 373 105
JP046837.1 - 3 - 4 373 105

View Full Product Details