Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gh1
Entrez Gene ID
100770534
Gene Name
growth hormone 1
Gene Aliases
I79_008312
Chromosome Location
Chr.Un|NW_003613864: 1699488 - 1701445 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642529.1 XP_007640719.1 1 - 2 222 84

View Full Product Details