Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gpr50
Entrez Gene ID
100772960
Gene Name
G protein-coupled receptor 50
Gene Aliases
I79_018289
Chromosome Location
Chr.Un|NW_003614816: 320188 - 325960 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007650696.1 XP_007648886.1 1 - 2 56 72

View Full Product Details