Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Art5
Entrez Gene ID
100759641
Gene Name
ADP-ribosyltransferase 5
Gene Aliases
I79_017914
Chromosome Location
Chr.Un|NW_003614758: 127161 - 129593 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509702.1 XP_003509750.1 3 - 4 825 64

View Full Product Details