Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hddc2
Entrez Gene ID
100753668
Gene Name
HD domain containing 2
Gene Aliases
I79_020595
Chromosome Location
Chr.Un|NW_003615289: 126380 - 141440 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652463.1 XP_007650653.1 3 - 4 433 82
GenBank mRNA JP038704.1 - 3 - 4 433 82

View Full Product Details