Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tet1
Entrez Gene ID
100771765
Gene Name
tet methylcytosine dioxygenase 1
Gene Aliases
I79_006066
Chromosome Location
Chr.Un|NW_003613759: 2104953 - 2167703 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499911.1 XP_003499959.1 3 - 4 4019 103

View Full Product Details