Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tsen15
Entrez Gene ID
100755550
Gene Name
TSEN15 tRNA splicing endonuclease subunit
Gene Aliases
I79_007140
Chromosome Location
Chr.Un|NW_003613816: 2497 - 15202 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500853.2 XP_003500901.2 3 - 4 518 109

View Full Product Details