Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam72a
Entrez Gene ID
100757979
Gene Name
family with sequence similarity 72, member A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613838: 1389427 - 1399130 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501214.2 XP_003501262.1 3 - 4 1190 86
GenBank mRNA JI871624.1 - 3 - 4 529 86

View Full Product Details