Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bmp6
Entrez Gene ID
100768517
Gene Name
bone morphogenetic protein 6
Gene Aliases
I79_008475
Chromosome Location
Chr.Un|NW_003613871: 1061083 - 1090784 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007642639.1 XP_007640829.1 6 - 7 775 76

View Full Product Details