Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Prox2
Entrez Gene ID
100758177
Gene Name
prospero homeobox 2
Gene Aliases
I79_009417
Chromosome Location
Chr.Un|NW_003613913: 18819 - 42149 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003502778.2 XP_003502826.1 2 - 3 1583 70

View Full Product Details