Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Aldoc
Entrez Gene ID
100760749
Gene Name
aldolase C, fructose-bisphosphate
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617063: 37316 - 41192 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003514876.2 XP_003514924.1 7 - 8 2799 81

View Full Product Details