Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdc3
Entrez Gene ID
100762010
Gene Name
syndecan 3
Gene Aliases
I79_004163, Syndecan-3
Chromosome Location
Chr.Un|NW_003613689: 728081 - 733813 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003498348.1 XP_003498396.1 3 - 4 1109 99

View Full Product Details