Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sfrp2
Entrez Gene ID
100771184
Gene Name
secreted frizzled-related protein 2
Gene Aliases
I79_005573
Chromosome Location
Chr.Un|NW_003613739: 1877231 - 1884406 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499474.2 XP_003499522.1 1 - 2 712 94

View Full Product Details