Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdkl4
Entrez Gene ID
100756171
Gene Name
cyclin-dependent kinase-like 4
Gene Aliases
I79_016413
Chromosome Location
Chr.Un|NW_003614547: 194293 - 227524 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003508459.2 XP_003508507.1 6 - 7 776 74

View Full Product Details