Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr86
Entrez Gene ID
100759512
Gene Name
WD repeat domain 86
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613842: 779731 - 796006 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501286.2 XP_003501334.1 4 - 5 856 106

View Full Product Details