Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tnfrsf13c
Entrez Gene ID
100762567
Gene Name
tumor necrosis factor receptor superfamily, member 13C
Gene Aliases
I79_021096
Chromosome Location
Chr.Un|NW_003615427: 178337 - 179537 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003512340.1 XP_003512388.1 1 - 2 143 92

View Full Product Details