Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Igflr1
Entrez Gene ID
100775057
Gene Name
IGF-like family receptor 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614167: 687803 - 689462 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007646071.1 XP_007644261.1 6 - 7 569 113

View Full Product Details