Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Otop3
Entrez Gene ID
100758515
Gene Name
otopetrin 3
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613598: 539848 - 546300 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643088.1 XP_007641278.1 3 - 4 597 61

View Full Product Details