Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ina
Entrez Gene ID
100763646
Gene Name
internexin neuronal intermediate filament protein, alpha
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003617470: 17812 - 26785 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007655167.1 XP_007653357.1 2 - 3 422 86

View Full Product Details