Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tcta
Entrez Gene ID
100763762
Gene Name
T-cell leukemia translocation altered
Gene Aliases
I79_006658
Chromosome Location
Chr.Un|NW_003613788: 1967175 - 1970014 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500356.2 XP_003500404.1 2 - 3 423 66

View Full Product Details