Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tceal5
Entrez Gene ID
100768643
Gene Name
transcription elongation factor A (SII)-like 5
Gene Aliases
I79_015642
Chromosome Location
Chr.Un|NW_003614444: 136122 - 136830 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648458.1 XP_007646648.1 1 - 1 521 90

View Full Product Details