Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Shisa9
Entrez Gene ID
100754470
Gene Name
shisa family member 9
Gene Aliases
I79_002156
Chromosome Location
Chr.Un|NW_003613631: 1957306 - 2254459 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652562.1 XP_007650752.1 3 - 4 716 82

View Full Product Details