Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Cygb
Entrez Gene ID
100762644
Gene Name
cytoglobin
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614300: 722686 - 732265 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506847.2 XP_003506895.1 4 - 5 644 68

View Full Product Details