Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rasa4b
Entrez Gene ID
100765808
Gene Name
RAS p21 protein activator 4B
Gene Aliases
Rasa4
Chromosome Location
Chr.Un|NW_003613822: 976530 - 993005 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641687.1 XP_007639877.1 15 - 16 1934 86

View Full Product Details