Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ino80b
Entrez Gene ID
100764793
Gene Name
INO80 complex subunit B
Gene Aliases
I79_019607
Chromosome Location
Chr.Un|NW_003615037: 281760 - 284861 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007651690.1 XP_007649880.1 5 - 6 804 111

View Full Product Details