Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clec12a
Entrez Gene ID
100769792
Gene Name
C-type lectin domain family 12, member A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614673: 98926 - 115577 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003509246.2 XP_003509294.1 4 - 5 527 85

View Full Product Details