Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LOC100764233
Entrez Gene ID
100764233
Gene Name
putative olfactory receptor GPCRLTM7
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003618362: 17770 - 18708 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003515534.1 XP_003515582.1 1 - 1 818 125

View Full Product Details