Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Faim3
Entrez Gene ID
100770414
Gene Name
Fas apoptotic inhibitory molecule 3
Gene Aliases
I79_003373
Chromosome Location
Chr.Un|NW_003613661: 2929873 - 2944969 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003497726.2 XP_003497774.1 6 - 7 1026 88

View Full Product Details