Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Epdr1
Entrez Gene ID
100770772
Gene Name
ependymin related 1
Gene Aliases
I79_001595
Chromosome Location
Chr.Un|NW_003613616: 59178 - 94445 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007648407.1 XP_007646597.1 3 - 4 435 95

View Full Product Details