Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hsbp1l1
Entrez Gene ID
100763672
Gene Name
heat shock factor binding protein 1-like 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613863: 488472 - 491194 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501844.1 XP_003501892.1 2 - 3 121 71

View Full Product Details