Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dclk3
Entrez Gene ID
100765735
Gene Name
doublecortin-like kinase 3
Gene Aliases
I79_014131
Chromosome Location
Chr.Un|NW_003614270: 678108 - 694894 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007647206.1 XP_007645396.1 1 - 2 1840 73

View Full Product Details