Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tnfsf14
Entrez Gene ID
100763671
Gene Name
tumor necrosis factor (ligand) superfamily, member 14
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613861: 1447253 - 1450882 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003501773.1 XP_003501821.1 3 - 4 284 82

View Full Product Details