Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Spz1
Entrez Gene ID
100760713
Gene Name
spermatogenic leucine zipper 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614442: 257770 - 258918 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003507869.1 XP_003507917.1 1 - 1 1017 89

View Full Product Details