Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Treml4
Entrez Gene ID
100757253
Gene Name
triggering receptor expressed on myeloid cells-like 4
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003615227: 20537 - 36809 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007652284.1 XP_007650474.1 6 - 7 1325 63
XM_007652285.1 XP_007650475.1 7 - 8 779 63
XM_007652286.1 XP_007650476.1 6 - 7 688 63
XM_007652287.1 XP_007650477.1 6 - 7 717 63
XM_007652288.1 XP_007650478.1 6 - 7 1237 63
GenBank mRNA JI878795.1 - 4 - 5 329 63

View Full Product Details