Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Klhl26
Entrez Gene ID
100756815
Gene Name
kelch-like family member 26
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613716: 765013 - 790314 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003499153.2 XP_003499201.1 3 - 4 354 85

View Full Product Details