Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ip6k1
Entrez Gene ID
100760280
Gene Name
inositol hexakisphosphate kinase 1
Gene Aliases
I79_006643
Chromosome Location
Chr.Un|NW_003613788: 1678724 - 1725585 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003500343.2 XP_003500391.1 2 - 3 575 83
GenBank mRNA JI873667.1 - 1 - 2 341 83
JP043196.1 - 1 - 2 342 83

View Full Product Details