Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Crygb
Entrez Gene ID
100759531
Gene Name
crystallin, gamma B
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003614305: 198411 - 200275 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003506913.2 XP_003506961.1 1 - 2 255 91

View Full Product Details