Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rasd1
Entrez Gene ID
100767666
Gene Name
RAS, dexamethasone-induced 1
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613960: 1256716 - 1260216 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007643975.1 XP_007642165.1 1 - 2 36 73

View Full Product Details