Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Syk
Entrez Gene ID
100758656
Gene Name
spleen tyrosine kinase
Gene Aliases
I79_010371, Sykb
Chromosome Location
Chr.Un|NW_003613980: 73964 - 119611 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007644151.1 XP_007642341.1 12 - 13 1814 66

View Full Product Details