Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Arl14
Entrez Gene ID
100757028
Gene Name
ADP-ribosylation factor-like 14
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613954: 1353767 - 1354604 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503264.2 XP_003503312.1 1 - 1 525 83

View Full Product Details