Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fgf16
Entrez Gene ID
100767099
Gene Name
fibroblast growth factor 16
Gene Aliases
I79_012477
Chromosome Location
Chr.Un|NW_003614139: 321277 - 331413 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003505303.2 XP_003505351.1 1 - 2 75 106

View Full Product Details