Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Rassf5
Entrez Gene ID
100758747
Gene Name
Ras association (RalGDS/AF-6) domain family member 5
Gene Aliases
I79_007609
Chromosome Location
Chr.Un|NW_003613838: 1660921 - 1731321 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007641912.1 XP_007640102.1 7 - 8 956 85

View Full Product Details