Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Maml2
Entrez Gene ID
100762611
Gene Name
mastermind-like 2 (Drosophila)
Gene Aliases
I79_006272
Chromosome Location
Chr.Un|NW_003613772: 719656 - 810865 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007640808.1 XP_007638998.1 2 - 3 1789 76
GenBank mRNA JP063271.1 - 1 - 2 221 76

View Full Product Details