Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ermn
Entrez Gene ID
100759143
Gene Name
ermin, ERM-like protein
Gene Aliases
Ermin
Chromosome Location
Chr.Un|NW_003613966: 1250498 - 1255762 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_003503410.1 XP_003503458.1 2 - 3 337 63

View Full Product Details