Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Kif26a
Entrez Gene ID
100750948
Gene Name
kinesin family member 26A
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613702: 1677131 - 1735240 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007633173.1 XP_007631363.1 12 - 13 1934 57

View Full Product Details