Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ndufa13
Entrez Gene ID
100762893
Gene Name
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 13
Gene Aliases
-
Chromosome Location
Chr.Un|NW_003613716: 213084 - 221143 on Build CriGri-PICRH-1.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_007638317.1 XP_007636507.1 2 - 3 133 62

View Full Product Details